siRNA Details
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107329.1 | NP_001100799.1 | 2 | 2401 | ||
XM_006253396.1 | XP_006253458.1 | 2 | 2392 | |||
GenBank mRNA | BC168662.1 | - | 2 | 2398 | 7930141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
