siRNA Details
Gene Symbol: Ctsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181378.2 | NP_852043.1 | 6 | 826 | ||
GenBank mRNA | AF456462.1 | - | 6 | 826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
