siRNA Details
Gene Symbol: Dapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107335.2 | NP_001100805.2 | 13, 14 | 1506 | ||
XM_006253522.2 | XP_006253584.1 | 12, 13 | 1349 | |||
XM_006253523.2 | XP_006253585.1 | 12, 13 | 1349 | |||
XM_008771440.1 | XP_008769662.1 | 12, 13 | 1349 | |||
XM_008771441.1 | XP_008769663.1 | 12, 13 | 1349 | |||
GenBank mRNA | BC169063.1 | - | 6, 7 | 563 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
