siRNA Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107336.1 | NP_001100806.1 | 5 | 1416 | ||
XM_006253619.2 | XP_006253681.1 | 5 | 1428 | |||
GenBank mRNA | BC166772.1 | - | 5 | 1408 | 7934143 |
