siRNA Details
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107337.1 | NP_001100807.1 | 5 | 1351 | ||
XM_006253620.2 | XP_006253682.1 | 5 | 1600 | |||
XM_006253621.2 | XP_006253683.1 | 5 | 1702 | |||
XM_006253622.2 | XP_006253684.1 | 5 | 1430 | |||
XM_006253623.2 | XP_006253685.1 | 5 | 1589 | |||
XM_006253624.2 | XP_006253686.1 | 5 | 1526 | |||
XM_006253625.2 | XP_006253687.1 | 5 | 1705 | |||
XM_006253627.2 | XP_006253689.1 | 5 | 1599 | |||
XM_008771491.1 | XP_008769713.1 | 5 | 1655 | |||
XM_008771492.1 | XP_008769714.1 | 4 | 566 | |||
XM_008771493.1 | XP_008769715.1 | 4 | 856 | |||
XM_008771494.1 | XP_008769716.1 | 5 | 1599 |
