siRNA Details
Gene Symbol: Arl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207165.1 | NP_997048.1 | 2 | 412 | ||
XM_006253629.1 | XP_006253691.1 | 2 | 460 | |||
GenBank mRNA | FQ210279.1 | - | 2 | 405 |

More Information
Other Species Targeted

Gene Ontology Categories:
