siRNA Details
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013138.2 | NP_001013156.2 | 4 | 502 | ||
XM_006253630.2 | XP_006253692.1 | 4 | 535 | |||
XM_006253631.2 | XP_006253693.1 | 3 | 390 | |||
GenBank mRNA | AY485933.1 | - | 4 | 497 | ||
BC097431.1 | - | 4 | 482 | 7455325 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
