siRNA Details
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013138.2 | NP_001013156.2 | 4 | 518 | ||
XM_006253630.2 | XP_006253692.1 | 4 | 551 | |||
XM_006253631.2 | XP_006253693.1 | 3 | 406 | |||
GenBank mRNA | AY485933.1 | - | 4 | 513 | ||
BC097431.1 | - | 4 | 498 | 7455325 | ||
FM038661.1 | - | 4 | 518 |
