siRNA Details
Gene Symbol: Iars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100572.1 | NP_001094042.1 | 18 | 2036 | ||
XM_006253706.2 | XP_006253768.1 | 18 | 1926 | |||
XM_008771533.1 | XP_008769755.1 | 19 | 2134 |
