siRNA Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008556.1 | NP_001008556.1 | 7, 8 | 558 | ||
XM_008771534.1 | XP_008769756.1 | 6, 7 | 1273 | |||
XM_008771535.1 | XP_008769757.1 | 7, 8 | 657 | |||
GenBank mRNA | AY823319.1 | - | 7, 8 | 558 |
