siRNA Details
Gene Symbol: Phf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107342.1 | NP_001100812.1 | 20 | 3048 | ||
XM_006253729.2 | XP_006253791.1 | 21 | 5070 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011078, BC051633, AK134358, AF043726, AB221590, and others.
Gene Ontology Categories:
