siRNA Details
Gene Symbol: Sirt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004256.1 | NP_001004256.1 | 4 | 564 | ||
XM_006253801.1 | XP_006253863.1 | 5 | 748 | |||
XM_006253802.1 | XP_006253864.1 | 5 | 626 | |||
XM_006253803.2 | XP_006253865.1 | 5 | 620 | |||
XM_006253804.2 | XP_006253866.1 | 5 | 571 | |||
GenBank mRNA | BC078958.1 | - | 4 | 564 | 7110841 |
