siRNA Details
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751664.3 | XP_003751712.1 | 2 | 201 | ||
XM_006253850.1 | XP_006253912.1 | 2 | 181 | |||
XM_006253852.2 | XP_006253914.1 | 2 | 341 | |||
XM_006253853.2 | XP_006253915.1 | 2 | 341 | |||
XM_006253854.2 | XP_006253916.1 | 2 | 341 | |||
XM_006253855.2 | XP_006253917.1 | 2 | 256 | |||
XM_006253856.2 | XP_006253918.1 | 2 | 188 | |||
XM_225253.8 | XP_225253.5 | 2 | 340 |
