siRNA Details
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012052.1 | NP_001012052.1 | 5 | 1583 | ||
XM_006253808.2 | XP_006253870.1 | 5 | 1570 | |||
XM_006253809.2 | XP_006253871.1 | 4 | 1261 | |||
XM_006253810.2 | XP_006253872.1 | 5 | 1600 | |||
XM_006253811.2 | XP_006253873.1 | 5 | 1684 | |||
GenBank mRNA | BC081934.1 | - | 5 | 1583 | 7114574 | |
FQ211932.1 | - | 5 | 1688 |
