siRNA Details
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107353.1 | NP_001100823.1 | 7 | 1104 | ||
XM_006253966.2 | XP_006254028.1 | 7 | 1181 | |||
XM_006253967.2 | XP_006254029.1 | 7 | 928 | |||
XM_006253968.2 | XP_006254030.1 | 7 | 1197 | |||
XM_006253969.2 | XP_006254031.1 | 7 | 1181 | |||
XM_008771654.1 | XP_008769876.1 | 7 | 2365 | |||
XM_008771655.1 | XP_008769877.1 | 7 | 2112 | |||
XM_008771656.1 | XP_008769878.1 | 7 | 2365 |
