siRNA Details
Gene Symbol: Zfp184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751710.3 | XP_003751758.1 | 6 | 861 | ||
XM_006254113.2 | XP_006254175.1 | 6 | 928 | |||
XM_006254114.2 | XP_006254176.1 | 6 | 1053 | |||
XM_006254118.2 | XP_006254180.1 | 3 | 784 | |||
XM_008771768.1 | XP_008769990.1 | 5 | 957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
