siRNA Details
Gene Symbol: Vps41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107355.1 | NP_001100825.1 | 18, 19 | 1565 | ||
GenBank mRNA | FQ211797.1 | - | 18, 19 | 1560 | ||
FQ221883.1 | - | 18, 19 | 1568 | |||
FQ223450.1 | - | 18, 19 | 1568 |
