siRNA Details
Gene Symbol: Yae1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107356.2 | NP_001100826.1 | 3 | 1432 | ||
GenBank mRNA | FQ211942.1 | - | 3 | 1438 | ||
FQ216975.1 | - | 3 | 1399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
