siRNA Details
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107357.1 | NP_001100827.1 | 17 | 2383 | ||
XM_006254011.2 | XP_006254073.1 | 17 | 2429 | |||
XM_006254012.2 | XP_006254074.1 | 16 | 2354 | |||
XM_006254013.2 | XP_006254075.1 | 16 | 2289 | |||
XM_008771727.1 | XP_008769949.1 | 15 | 2214 | |||
XM_008771728.1 | XP_008769950.1 | 14 | 1459 |
