siRNA Details
Gene Symbol: Arhgap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107357.1 | NP_001100827.1 | 3 | 344 | ||
XM_006254011.2 | XP_006254073.1 | 3 | 391 | |||
XM_006254012.2 | XP_006254074.1 | 3 | 391 | |||
XM_006254013.2 | XP_006254075.1 | 3 | 392 | |||
XM_008771727.1 | XP_008769949.1 | 3 | 392 |
