siRNA Details
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270417.1 | NP_001257346.1 | 12 | 1784 | ||
GenBank mRNA | AB574169.1 | - | 12 | 1784 |

More Information
Other Species Targeted

Gene Ontology Categories:
