siRNA Details
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012468.1 | NP_001012486.1 | 3 | 208 | ||
XM_006254132.2 | XP_006254194.1 | 3 | 265 | |||
GenBank mRNA | BC089957.1 | - | 3 | 208 | 7312864 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_181070, NM_001278447, X80333, AK143673, L04966, and others.
Gene Ontology Categories:
