siRNA Details
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107362.1 | NP_001100832.1 | 8 | 909 | ||
XM_008771734.1 | XP_008769956.1 | 9 | 922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
