siRNA Details
Gene Symbol: Akr1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033697.1 | NP_001028869.1 | 4 | 395 | ||
XM_006254188.2 | XP_006254250.1 | 4 | 449 | |||
GenBank mRNA | AB300410.1 | - | 4 | 386 | ||
BC088227.1 | - | 4 | 384 | |||
BC104716.1 | - | 4 | 395 | 7373256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
