siRNA Details
Gene Symbol: Txndc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024998.1 | NP_001020169.1 | 4, 5 | 902 | ||
XM_008771497.1 | XP_008769719.1 | 4, 5 | 1010 | |||
GenBank mRNA | BC091340.1 | - | 4, 5 | 902 | 7315216 |
