siRNA Details
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007687.1 | NP_001007688.1 | 10 | 1402 | ||
XM_006255009.2 | XP_006255071.1 | 11 | 3124 | |||
XM_006255010.2 | XP_006255072.1 | 10 | 2960 | |||
XM_008772135.1 | XP_008770357.1 | 11 | 1424 | |||
GenBank mRNA | BC081877.1 | - | 10 | 1402 | 7107931 |
