siRNA Details
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271281.1 | NP_001258210.1 | 10 | 852 | ||
XM_008772138.1 | XP_008770360.1 | 9 | 1105 | |||
XM_008772139.1 | XP_008770361.1 | 9 | 1105 | |||
GenBank mRNA | CB770049.1 | - | 6 | 355 | ||
CK364772.1 | - | 2 | 77 |
