siRNA Details
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083687.1 | - | 2 | 565 | 7189262 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083687.1 | - | 2 | 565 | 7189262 |