siRNA Details
Gene Symbol: Zfp532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107382.1 | NP_001100852.1 | 2, 3 | 157 | ||
XM_006254865.2 | XP_006254927.1 | 2, 3 | 2447 | |||
XM_006254866.2 | XP_006254928.1 | 2, 3 | 1886 | |||
XM_006254868.2 | XP_006254930.1 | 2, 3 | 474 | |||
XM_006254869.2 | XP_006254931.1 | 2, 3 | 319 | |||
XM_006254870.2 | XP_006254932.1 | 2, 3 | 1851 | |||
XM_006254875.2 | XP_006254937.1 | 1, 2 | 1073 | |||
XM_006254876.2 | XP_006254938.1 | 1, 2 | 1073 | |||
XM_008772142.1 | XP_008770364.1 | 1, 2 | 1073 |
