siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254839.2 | XP_006254901.1 | 35, 36 | 5902 | ||
XM_225880.8 | XP_225880.5 | 34, 35 | 5858 |

More Information
Other Species Targeted

Gene Ontology Categories:
