siRNA Details
Gene Symbol: Sh3tc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254817.2 | XP_006254879.1 | 11 | 2993 | ||
XM_006254818.2 | XP_006254880.1 | 11 | 2872 | |||
XM_008772199.1 | XP_008770421.1 | 11 | 2992 | |||
XM_008772200.1 | XP_008770422.1 | 11 | 2992 | |||
XM_008772201.1 | XP_008770423.1 | 11 | 2992 | |||
XM_008772202.1 | XP_008770424.1 | 8 | 2586 | |||
XM_225887.8 | XP_225887.5 | 11 | 2992 |

More Information
Other Species Targeted

Gene Ontology Categories:
