siRNA Details
Gene Symbol: Sh3tc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254817.2 | XP_006254879.1 | 17 | 4494 | ||
XM_006254818.2 | XP_006254880.1 | 17 | 4373 | |||
XM_008772202.1 | XP_008770424.1 | 14 | 4087 | |||
XM_225887.8 | XP_225887.5 | 17 | 3863 | |||
GenBank mRNA | BC089936.1 | - | 3 | 267 |
