siRNA Details
Gene Symbol: Pcyox1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134542.1 | NP_001128014.1 | 2 | 193 | ||
XM_006254806.2 | XP_006254868.1 | 2 | 849 | |||
GenBank mRNA | BC168676.1 | - | 2 | 193 | 5598155 |

More Information
Other Species Targeted

Gene Ontology Categories:
