siRNA Details
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100974.1 | NP_001094444.1 | 4 | 449 | ||
XM_006254807.2 | XP_006254869.1 | 4 | 508 | |||
XM_006254808.2 | XP_006254870.1 | 4 | 2448 | |||
XM_006254809.2 | XP_006254871.1 | 4 | 415 | |||
XM_006254811.2 | XP_006254873.1 | 4 | 544 | |||
GenBank mRNA | AB199794.1 | - | 4 | 449 | ||
BC168236.1 | - | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
