siRNA Details
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751784.3 | XP_003751832.1 | 18 | 3370 | ||
XM_006254710.2 | XP_006254772.1 | 18 | 3370 | |||
XM_006254711.2 | XP_006254773.1 | 18 | 3370 | |||
XM_006254712.2 | XP_006254774.1 | 18 | 3370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
