siRNA Details
Gene Symbol: RGD735029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199493.1 | NP_955787.1 | 2, 3 | 181 | ||
XM_006254634.2 | XP_006254696.1 | 2, 3 | 288 | |||
GenBank mRNA | BC064660.1 | - | 2, 3 | 181 | 5623815 | |
FQ211991.1 | - | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
