siRNA Details
Gene Symbol: AXIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004655.3 | NP_004646.3 | 7 | 2024 | ||
XM_011525319.2 | XP_011523621.1 | 7 | 2235 | |||
XM_011525320.1 | XP_011523622.1 | 7 | 2224 | |||
XM_011525321.2 | XP_011523623.1 | 7 | 2172 | |||
XM_017025192.1 | XP_016880681.1 | 7 | 1891 | |||
GenBank mRNA | AF078165.1 | - | 6 | 1824 | ||
AK025718.1 | - | 6 | 1046 | |||
AK315303.1 | - | 7 | 1940 | |||
BC101533.1 | - | 6 | 1804 | 8069039 | ||
CR933657.1 | - | 7 | 2049 | |||
DA839648.1 | - | 2 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
