siRNA Details
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025414.1 | NP_001020585.1 | 10 | 1174 | ||
GenBank mRNA | BC097969.1 | - | 10 | 1174 | 7442005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
