siRNA Details
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017932.1 | NP_001017932.1 | 5 | 718 | ||
GenBank mRNA | BC127460.1 | - | 5 | 673 | 8367400 |

More Information
Other Species Targeted

Gene Ontology Categories:
