siRNA Details
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008344.1 | NP_001008345.1 | 10 | 1240 | ||
GenBank mRNA | BC085902.1 | - | 10 | 1240 | 7193574 | |
FQ227237.1 | - | 10 | 1269 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144866, AK169345, AK077559, AK164020, AK049607, and others.
Gene Ontology Categories:
