siRNA Details
Gene Symbol: Epb41l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107397.2 | NP_001100867.1 | 12 | 1018 | ||
XM_006254582.2 | XP_006254644.1 | 17 | 4290 | |||
XM_006254583.2 | XP_006254645.1 | 17 | 3396 | |||
XM_006254584.2 | XP_006254646.1 | 18 | 3042 | |||
XM_006254588.2 | XP_006254650.1 | 17 | 1596 | |||
XM_008772040.1 | XP_008770262.1 | 17 | 1373 | |||
GenBank mRNA | BC168858.1 | - | 12 | 1018 | 7097125 |
