siRNA Details
Gene Symbol: Wdr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107398.2 | NP_001100868.2 | 5, 6 | 630 | ||
XM_006254589.2 | XP_006254651.1 | 5, 6 | 697 | |||
XM_008772041.1 | XP_008770263.1 | 5, 6 | 679 |

More Information
Other Species Targeted

Gene Ontology Categories:
