siRNA Details
Gene Symbol: Zfp521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107403.1 | NP_001100873.1 | 3 | 1461 | ||
XM_008771946.1 | XP_008770168.1 | 3 | 1589 | |||
XM_008771947.1 | XP_008770169.1 | 3 | 1589 | |||
XM_008771948.1 | XP_008770170.1 | 3 | 1567 | |||
XM_008771949.1 | XP_008770171.1 | 3 | 1589 |

More Information
Other Species Targeted

Gene Ontology Categories:
