siRNA Details
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772250.1 | XP_008770472.1 | 12 | 1631 | ||
XM_008772251.1 | XP_008770473.1 | 12 | 1552 | |||
XM_008772252.1 | XP_008770474.1 | 12 | 1839 | |||
XM_008772253.1 | XP_008770475.1 | 11 | 1710 | |||
XM_008772254.1 | XP_008770476.1 | 12 | 1842 | |||
XM_008772255.1 | XP_008770477.1 | 11 | 1689 | |||
XM_008772256.1 | XP_008770478.1 | 11 | 1686 | |||
XM_008772257.1 | XP_008770479.1 | 12 | 1842 |
