siRNA Details
Gene Symbol: Rpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246606.2 | XP_006246668.1 | 3 | 406 | ||
GenBank mRNA | FQ209942.1 | - | 3 | 252 | ||
FQ210129.1 | - | 3 | 252 | |||
FQ210265.1 | - | 3 | 252 | |||
FQ210305.1 | - | 3 | 253 | |||
FQ211391.1 | - | 3 | 275 | |||
FQ214577.1 | - | 3 | 252 | |||
FQ218083.1 | - | 3 | 252 | |||
FQ218115.1 | - | 3 | 251 | |||
FQ221223.1 | - | 3 | 254 | |||
FQ221302.1 | - | 3 | 252 | |||
FQ221303.1 | - | 3 | 252 | |||
FQ221304.1 | - | 3 | 252 | |||
FQ221369.1 | - | 3 | 252 | |||
FQ221596.1 | - | 3 | 247 | |||
FQ221712.1 | - | 3 | 252 | |||
FQ221762.1 | - | 3 | 252 | |||
FQ222089.1 | - | 3 | 252 | |||
FQ222135.1 | - | 3 | 252 | |||
FQ222176.1 | - | 3 | 252 | |||
FQ222323.1 | - | 3 | 246 | |||
FQ222676.1 | - | 3 | 252 | |||
FQ222872.1 | - | 3 | 252 | |||
FQ222877.1 | - | 3 | 246 | |||
FQ222953.1 | - | 3 | 252 | |||
FQ223076.1 | - | 3 | 253 | |||
FQ223161.1 | - | 3 | 252 | |||
FQ224611.1 | - | 3 | 252 | |||
FQ228453.1 | - | 3 | 251 | |||
FQ228549.1 | - | 3 | 246 | |||
FQ228592.1 | - | 3 | 246 | |||
FQ228636.1 | - | 3 | 252 | |||
FQ228935.1 | - | 3 | 253 | |||
X14671.1 | - | 3 | 247 |
Gene Symbol: LOC100910721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751828.3 | XP_003751876.1 | 1 | 211 |
