siRNA Details
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107413.1 | NP_001100883.1 | 1 | 496 | ||
XM_006255191.2 | XP_006255253.1 | 2 | 1475 | |||
XM_006255192.2 | XP_006255254.1 | 2 | 1475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
