siRNA Details
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107413.1 | NP_001100883.1 | 1 | 583 | ||
XM_006255191.2 | XP_006255253.1 | 2 | 1562 | |||
XM_006255192.2 | XP_006255254.1 | 2 | 1562 |
