siRNA Details
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107416.1 | NP_001100886.1 | 2 | 306 | ||
XM_006255261.1 | XP_006255323.1 | 2 | 712 | |||
XM_006255263.2 | XP_006255325.1 | 2 | 712 | |||
XM_008772403.1 | XP_008770625.1 | 3 | 723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
