siRNA Details
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107416.1 | NP_001100886.1 | 8 | 999 | ||
XM_006255261.1 | XP_006255323.1 | 10 | 1609 | |||
XM_006255263.2 | XP_006255325.1 | 10 | 1609 | |||
XM_008772403.1 | XP_008770625.1 | 11 | 1620 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001164498, NM_001164499, NM_001164497, BC145737, BC144796, and others.
Gene Ontology Categories:
