siRNA Details
Gene Symbol: Neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107417.2 | NP_001100887.2 | 3 | 453 | ||
GenBank mRNA | GQ149762.1 | - | 3 | 453 |

More Information
Other Species Targeted

Gene Ontology Categories:
